Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Ephrin-A2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Ephrin-A2 |
---|---|
Clone | 84019 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Ephrin-A2 Monoclonal specifically detects Ephrin-A2 in Mouse samples. It is validated for Western Blot.Specifications
Ephrin-A2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cek7-L, EFNA2, ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EphrinA2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6, LERK6LERK-6, ligand of eph-related kinase 6 | |
EFNA2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
84019 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P52801 | |
1943 | |
Mouse myeloma cell line NS0-derived recombinant mouse Ephrin-A2 Arg21-Asn184 Accession # P52801 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Ephrin-A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Ephrin-A4, recombinant human (rh) Ephrin-A3, rmEphrin-B1, and rhEphrin-B3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title