Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse FGFR5/FGFRL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064702
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1899SP 25 μg
MAB1899 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1899SP Supplier R&D Systems Supplier No. MAB1899SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen FGFR5/FGFRL1
Applications Western Blot
Classification Monoclonal
Clone 223003
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q91V87
Gene Alias FGF homologous factor receptor, FGF R5, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFRL1, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Gene Symbols Fgfrl1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse FGF R5/FGFRL1 Ala21-Pro374 Accession # Q91V87
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53834
Test Specificity Detects mouse FGF R5/FGFRL1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), 1 alpha (IIIc), 1 beta (IIIb), 1 beta (IIIc), recombinant mouse (rm) FGF R2 alpha (IIIb), 2 alpha (IIIc), 2 beta (IIIb), 2 beta (IIIc), 3 alpha (IIIb), 3 alpha (IIIc), rmFGF R4, or rhFGF R5 is observed. Was not tested for cross-reactivity with rmFGF R5 gamma .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.