Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Fibroblast Activation Protein alpha/FAP Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.16 - $579.52
Specifications
| Antigen | Fibroblast Activation Protein alpha/FAP |
|---|---|
| Clone | 983802 |
| Dilution | ELISA, Immunocytochemistry 8-25 ug/mL |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9727SP
![]() |
R&D Systems
MAB9727SP |
25 μg |
Each for $200.16
|
|
|||||
MAB9727100
![]() |
R&D Systems
MAB9727100 |
100 μg |
Each for $579.52
|
|
|||||
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry.Specifications
| Fibroblast Activation Protein alpha/FAP | |
| ELISA, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P97321 | |
| 2191 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 983802 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fibroblast Activation Protein alpha/FAP Leu26-Asp761 Accession # P97321 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title