Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1677
This item is not returnable.
View return policy
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Immunohistochemistry.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GFRA4 | |
Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 | |
500 μg | |
Primary | |
Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse GFR alpha-2, recombinant human (rh) GFR alpha-3, recombinant rat GFR alpha-1, or rhGFR alpha-4 is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
171217 | |
Immunohistochemistry 8-25 ug/mL | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64096 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction