Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
---|---|
Clone | 171217 |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Immunohistochemistry.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
64096 | |
Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse GFR alpha-2, recombinant human (rh) GFR alpha-3, recombinant rat GFR alpha-1, or rhGFR alpha-4 is observed. |
171217 | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
GFRA4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title