Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-like Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB5728100
This item is not returnable.
View return policy
Description
GFR alpha-like Monoclonal antibody specifically detects GFR alpha-like in Mouse samples. It is validated for ImmunocytochemistrySpecifications
GFR alpha-like | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Immunocytochemistry | |
1045708 | |
Immunocytochemistry 8-25 ug/mL | |
Q6SJE0-1 | |
Chinese Hamster Ovary cell line CHO-derived mouse GFR alpha-like, Gln20-Gly349, Accession # Q6SJE0-1 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction