Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GFR alpha-like Antibody, R&D Systems™

Rat Monoclonal Antibody
$139.50 - $409.00
Specifications
Antigen | GFR alpha-like |
---|---|
Clone | 1045708 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5728SP
![]() |
R&D Systems
MAB5728SP |
25 μg |
Each for $139.50
|
|
|||||
MAB5728100
![]() |
R&D Systems
MAB5728100 |
100 μg |
Each for $409.00
|
|
|||||
Description
GFR alpha-like Monoclonal antibody specifically detects GFR alpha-like in Mouse samples. It is validated for ImmunocytochemistrySpecifications
GFR alpha-like | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
Chinese Hamster Ovary cell line CHO-derived mouse GFR alpha-like, Gln20-Gly349, Accession # Q6SJE0-1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1045708 | |
Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q6SJE0-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title