Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse HSF4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$176.44 - $532.55
Specifications
| Antigen | HSF4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HSF4 Polyclonal specifically detects HSF4 in Mouse samples. It is validated for Western Blot.Specifications
| HSF4 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| ATHSF4, AT-HSFB1, cataract, Marner, CTM, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSFB1, HSTF 4 | |
| HSF4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9R0L1 | |
| 3299 | |
| E. coli-derived recombinant mouse HSF4 Gly343-Pro492 Accession # Q9R0L1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse HSF4 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) HSF4 is observed and less than 5% cross-reactivity with recombinant mouse HSF1 and rhHSF2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title