Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgE Antibody (LO-ME-3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB310111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310111 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310111 Supplier Novus Biologicals Supplier No. NBP142334AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Mouse IgE Monoclonal antibody specifically detects Mouse IgE in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen Mouse IgE
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LO-ME-3
Conjugate Alexa Fluor 350
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias Immunoglobulin E, ImmunoglobulinE
Host Species Rat
Immunogen Mouse IgE
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.