Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGF-II/IGF2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB792SP
This item is not returnable.
View return policy
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGF-II | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3481 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, ELISA | |
122404 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P09535 | |
IGF2 | |
E. coli-derived recombinant mouse IGF-II Ala25-Glu91 Accession # P09535 | |
25 μg | |
Primary | |
Detects mouse IGF-II in ELISAs and Western blots. In ELISAs, this antibody shows less than 6% cross-reactivity with recombinant human (rh) IGF-I R and no cross-reactivity with rhIGF-I and rmIGF-I is observed. In Western blots, 100% cross-reactivity with rhIGF-II and no cross reactivity with rmIGF-I is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction