Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGF-II/IGF2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$200.50 - $1973.00
Specifications
Antigen | IGF-II |
---|---|
Clone | 122404 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
IGF-II | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
IGF2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
122404 | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P09535 | |
3481 | |
E. coli-derived recombinant mouse IGF-II Ala25-Glu91 Accession # P09535 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IGF-II in ELISAs and Western blots. In ELISAs, this antibody shows less than 6% cross-reactivity with recombinant human (rh) IGF-I R and no cross-reactivity with rhIGF-I and rmIGF-I is observed. In Western blots, 100% cross-reactivity with rhIGF-II and no cross reactivity with rmIGF-I is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title