Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-L1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4130SP
This item is not returnable.
View return policy
Description
IGFBP-L1 Polyclonal specifically detects IGFBP-L1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IGFBP-L1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA113O24.1, IGFBPL1, IGFBPL1 insulin-like growth factor binding protein-like 1, IGFBP-RP4 | |
Goat | |
Affinity Purified | |
RUO | |
347252 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q80W15 | |
IGFBPL1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-L1 Leu18-Leu270 Accession # Q80W15 | |
25 μg | |
Primary | |
Detects mouse IGFBP-L1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IGFBP-L1 is observed and less than 1% cross-reactivity with recombinant mouse IGFBP-7 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction