Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-L1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | IGFBP-L1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
IGFBP-L1 Polyclonal specifically detects IGFBP-L1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IGFBP-L1 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
bA113O24.1, IGFBPL1, IGFBPL1 insulin-like growth factor binding protein-like 1, IGFBP-RP4 | |
IGFBPL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q80W15 | |
347252 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-L1 Leu18-Leu270 Accession # Q80W15 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IGFBP-L1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human IGFBP-L1 is observed and less than 1% cross-reactivity with recombinant mouse IGFBP-7 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title