Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse IL-17 DuoSet ELISA
Click to view available options
Packaging Quantity:
15 Plates
5 Plates
Description
The Mouse IL-17 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-17/IL-17A.
Specifications
Specifications
Product Type | ELISA Development Kit |
Quantity | 1 Kit |
Species | Mouse |
Conjugate | Biotin |
Assay Range | 15.60 to 1000 pg/mL |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 16171 |
Synonym | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
Packaging Quantity | 15 Plates |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction