Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RB Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
$221.50 - $456.00
Specifications
| Antigen | IL-17RB |
|---|---|
| Clone | 752101 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB10402G02
![]() |
R&D Systems
FAB10402G025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB10402G10
![]() |
R&D Systems
FAB10402G100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
IL-17RB Monoclonal specifically detects IL-17RB in Mouse samples. It is validated for Flow Cytometry.Specifications
| IL-17RB | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
| IL17RB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse IL-17 RB in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 RB, rhIL-17 R, or recombinant mouse IL-17 is observed. |
| 752101 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| Mouse | |
| Q9JIP3 | |
| 55540 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RB Arg18-Gly286 Accession # Q9JIP3 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title