Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RE Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7997SP
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| IL-17RE | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 132014 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 944904 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q8BH06 | |
| Il17re | |
| Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE Met1-His414 Accession # Q8BH06 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-17 RE in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction