Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-17RE Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $538.00
Specifications
| Antigen | IL-17RE |
|---|---|
| Clone | 944904 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
Description
IL-17RE Monoclonal specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| IL-17RE | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E | |
| Il17re | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 944904 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8BH06 | |
| 132014 | |
| Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE Met1-His414 Accession # Q8BH06 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-17 RE in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title