Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-21R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5961SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5961SP 25 μg
MAB5961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5961SP Supplier R&D Systems Supplier No. MAB5961SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-21R Monoclonal specifically detects IL-21R in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IL-21R
Applications Western Blot
Classification Monoclonal
Clone 155516
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9JHX3
Gene Alias CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Gene Symbols IL21R
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-21 R Cys20-Pro236 Accession # Q9JHX3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50615
Test Specificity Detects mouse IL-21 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IL-21 R or recombinant mouse IL-2 R beta is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.