Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Integrin alpha 10 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Integrin alpha 10 |
---|---|
Clone | 885501 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9757SP
![]() |
R&D Systems
MAB9757SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9757100
![]() |
R&D Systems
MAB9757100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Integrin alpha 10 Monoclonal specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Integrin alpha 10 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Mouse | |
NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) | |
8515 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
885501 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
integrin alpha-10, integrin, alpha 10, ITGA10, PRO827 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1 Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1) Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Integrin alpha 10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Intagrin alpha 2 beta 1 was observered. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title