Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Latent TGF-beta bp4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
$200.50 - $549.50
Specifications
Antigen | Latent TGF-beta bp4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Latent TGF-beta bp4 Polyclonal specifically detects Latent TGF-beta bp4 in Mouse samples. It is validated for Western Blot.Specifications
Latent TGF-beta bp4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Latent TGFbeta bp4, latent transforming growth factor beta binding protein 4, LTBP4, LTBP-4, LTBP4L, LTBP4S | |
LTBP4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAI41006 | |
8425 | |
Mouse myeloma cell line NS0-derived recombinant mouse Latent TGF-beta bp4 isoform 1 Ser25-Ala1666 (Arg542-Pro547) Accession # AAI41006 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Latent TGF-beta bp4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) LTBP-1 and rhLTBP-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title