Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Lgr5/GPR49 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7121504
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB82401PL 25 μg
FAB82401PP 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB82401PL Supplier R&D Systems Supplier No. FAB82401P025UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Lgr5/GPR49
Applications Flow Cytometry
Classification Monoclonal
Clone 889901
Conjugate R-PE
Dilution Flow Cytometry 0.1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9Z1P4
Gene Alias FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Rat
Immunogen Mouse Lgr5/GPR49 N-terminal peptide Ala21-Cys34 Accession # Q9Z1P4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8549
Test Specificity Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5 transfected cells but not irrelevant transfectants in Flow Cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.