Learn More
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Based on its helical structure, LIF (Leukemia Inhibitory Factor) is considered a member of the Interleukin-6 family of cytokines. Functionally, it has been implicated in a many physiological processes including development, hematopoiesis, bone metabolism, and inflammation. Some cell types known to express LIF include activated T cells, monocytes, astrocytes, osteoblasts, keratinocytes, regenerating skeletal muscle, mast cells, and fibroblasts. The activities of LIF are mediated through a high-affinity heterodimeric receptor complex consisting of two membrane glycoproteins: an alpha subunit (LIF R alpha, also known as LIF R beta and CD118) that binds LIF with low affinity and the 130 kDa (gp130) subunit that does not bind LIF by itself, but is required for high-affinity binding of LIF by the complex.
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit |
| Species | Mouse |
| Conjugate | Biotin |
| Binding Target | LIF |
| Assay Range | 6.25 to 400 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3976 |
| Product Line | R&D Systems |
| Synonym | CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
