Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LILRC1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $538.00
Specifications
Antigen | LILRC1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
LILRC1 Polyclonal specifically detects LILRC1 in Mouse samples. It is validated for Western Blot.Specifications
LILRC1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
232801 | |
Mouse myeloma cell line NS0-derived recombinant mouse LILRC1 Gln17-Asn248 Accession # AAY44811 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse LILRC1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant rat (rr) LILRC1 is observed and approximately 15% cross-reactivity with rrLILRC2 is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAY44811 | |
Lilra5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title