Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lipin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4064
This item is not returnable.
View return policy
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Lipin 3 | |
Polyclonal | |
Unconjugated | |
Q99PI4 | |
LPIN3 | |
E. coli-derived recombinant mouse Lipin 3 Thr428-Arg529 Accession # Q99PI4 | |
100 μg | |
Primary | |
Detects mouse Lipin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Lipin 3 is observed and less than 5% cross-reactivity with rhLipin 1 and rhLipin 2 is observed. | |
Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, LPIN3, phosphatidate phosphatase LPIN3, SMP2 | |
Goat | |
Affinity Purified | |
RUO | |
64900 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction