Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lipin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Lipin 3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Lipin 3 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, LPIN3, phosphatidate phosphatase LPIN3, SMP2 | |
LPIN3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q99PI4 | |
64900 | |
E. coli-derived recombinant mouse Lipin 3 Thr428-Arg529 Accession # Q99PI4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Lipin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Lipin 3 is observed and less than 5% cross-reactivity with rhLipin 1 and rhLipin 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title