Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LRIG1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3688SP
This item is not returnable.
View return policy
Description
LRIG1 Monoclonal specifically detects LRIG1 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
LRIG1 | |
Monoclonal | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
P70193 | |
Lrig1 | |
Mouse myeloma cell line NS0-derived recombinant mouse LRIG1 Ala37-Thr794 Accession # P70193 | |
25 μg | |
Primary | |
Detects mouse LRIG1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LRIG3 is observed. | |
Mouse | |
Purified |
Blockade of Receptor-ligand Interaction | |
440710 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D6Bwg0781e, Img, LIG1, LIG-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26018 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction