Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LRIG1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | LRIG1 |
---|---|
Clone | 440710 |
Concentration | LYOPH |
Dilution | Blockade of Receptor-ligand Interaction |
Applications | Blockade of Receptor-ligand Interaction |
Description
LRIG1 Monoclonal specifically detects LRIG1 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
LRIG1 | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Rat | |
Mouse | |
P70193 | |
26018 | |
Mouse myeloma cell line NS0-derived recombinant mouse LRIG1 Ala37-Thr794 Accession # P70193 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse LRIG1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LRIG3 is observed. |
440710 | |
Blockade of Receptor-ligand Interaction | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D6Bwg0781e, Img, LIG1, LIG-1 | |
Lrig1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title