Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse LRP-6 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | LRP-6 |
---|---|
Clone | 290913 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
LRP-6 Monoclonal specifically detects LRP-6 in Mouse samples. It is validated for Western Blot.Specifications
LRP-6 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O88572 | |
4040 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse LRP-6 Met1-Pro1368 Accession # O88572 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse LRP-6 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 50 - 60% cross-reactivity with recombinant human (rh) LRP-6 and no cross-reactivity with rhLDL R. |
290913 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP6, LRP-6 | |
LRP6 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title