Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MAGP-1/MFAP2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4977
This item is not returnable.
View return policy
Description
MAGP-1/MFAP2 Polyclonal specifically detects MAGP-1/MFAP2 in Mouse samples. It is validated for Western Blot.Specifications
| MAGP-1/MFAP2 | |
| Polyclonal | |
| Unconjugated | |
| P55002 | |
| MFAP2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MAGP-1/MFAP2 Gln17-Cys183 Accession # P55002 | |
| 100 μg | |
| Primary | |
| Detects mouse MAGP-1/MFAP2 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human MAGP-1 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| MAGP1, MAGP-1FLJ50901, MAGPMFAP-2, MFAP2, Microfibril-associated glycoprotein 1, microfibrillar-associated protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4237 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction