Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MAGP-1/MFAP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7062571
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4977SP 25 μg
AF4977 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4977SP Supplier R&D Systems Supplier No. AF4977SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

MAGP-1/MFAP2 Polyclonal specifically detects MAGP-1/MFAP2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen MAGP-1/MFAP2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P55002
Gene Alias MAGP1, MAGP-1FLJ50901, MAGPMFAP-2, MFAP2, Microfibril-associated glycoprotein 1, microfibrillar-associated protein 2
Gene Symbols MFAP2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MAGP-1/MFAP2 Gln17-Cys183 Accession # P55002
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4237
Test Specificity Detects mouse MAGP-1/MFAP2 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human MAGP-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.