Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mast Cell Protease-1/Mcpt1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Mast Cell Protease-1/Mcpt1 |
---|---|
Clone | 285008 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Simple Western 10 ug/mL |
Applications | Western Blot |
Description
Mast Cell Protease-1/Mcpt1 Monoclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for Western Blot, Simple Western.Specifications
Mast Cell Protease-1/Mcpt1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse, Rat | |
P11034 | |
17224 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-1/Mcpt1 Ile21-Lys246 Accession # P11034 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Mast Cell Protease-1/Mcpt1 in direct ELISAs and Western blots. Detects rat Mast Cell Protease-1/Mcpt1 in Simple Western. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCP-6, -7, or -11 is observed. |
285008 | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AV080368, Mast Cell Protease1, Mcp1, Mcp-1, MMPC-1 | |
Mcpt1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title