Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MBL-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2077SP
This item is not returnable.
View return policy
Description
MBL1 Polyclonal specifically detects MBL1 in Mouse samples. It is validated for Western Blot.Specifications
MBL1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MBL1, MBL-1, MBL-A, MBP-A, p28B, RaRF p28B, S-MBP | |
Goat | |
Affinity Purified | |
RUO | |
17194 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
P39039 | |
Mbl1 | |
Mouse myeloma cell line NS0-derived recombinant mouse MBL-1 (R&D Systems, Catalog # 2077-MB) Ser18-Ala239 Accession # P39039 | |
25 μg | |
Primary | |
Detects mouse MBL-1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human MBL and recombinant mouse MBL-2 is observed under reducing conditions. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction