Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MBL-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | MBL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
MBL1 Polyclonal specifically detects MBL1 in Mouse samples. It is validated for Western Blot.Specifications
MBL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
MBL1, MBL-1, MBL-A, MBP-A, p28B, RaRF p28B, S-MBP | |
Mbl1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P39039 | |
17194 | |
Mouse myeloma cell line NS0-derived recombinant mouse MBL-1 (R&D Systems, Catalog # 2077-MB) Ser18-Ala239 Accession # P39039 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse MBL-1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human MBL and recombinant mouse MBL-2 is observed under reducing conditions. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title