Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MDL-1/CLEC5A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1639SP
This item is not returnable.
View return policy
Description
MDL-1/CLEC5A Monoclonal specifically detects MDL-1/CLEC5A in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
MDL-1/CLEC5A | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CLEC5A, CLECSF5, CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL1, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23601 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
226402 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CLEC5A | |
BaF3 mouse pro-B cell line transfected with mouse MDL-1/CLEC5A | |
25 μg | |
Primary | |
Detects mouse MDL-1/CLEC5A. In ELISAs, approximately 50% cross-reactivity with recombinant human MDL-1 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction