Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MDL-1/CLEC5A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$200.50 - $655.00
Specifications
Antigen | MDL-1/CLEC5A |
---|---|
Clone | 226402 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
MDL-1/CLEC5A Monoclonal specifically detects MDL-1/CLEC5A in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
MDL-1/CLEC5A | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
CLEC5A, CLECSF5, CLECSF5carbohydrate-recognition domain) lectin, superfamilymember 5, C-type lectin domain family 5, member A, C-type lectin superfamily member 5, MDL1, MDL-1MGC138304, Myeloid DAP12-associating lectin 1, myeloid DAP12-associating lectin-1 | |
CLEC5A | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
226402 | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
23601 | |
BaF3 mouse pro-B cell line transfected with mouse MDL-1/CLEC5A | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse MDL-1/CLEC5A. In ELISAs, approximately 50% cross-reactivity with recombinant human MDL-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title