Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse MUP-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB170577 Shop All Novus Biologicals Products
Click to view available options
Quantity:
1 Kit

Colorimetric ELISA Kit

Specifications

Assay Range 0.125 - 8 ng/mL (example only; lot dependent)
Assay Sensitivity 79 pg/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture, Plasma, Serum, Urine
For Use With (Application) ELISA
Gene ID (Entrez) 17840
Gene Symbol MUP1
Quantity 1 Kit
Research Discipline metabolism
Storage Requirements Storage is content dependent.
Synonym Ltn-1, Lvtn-1, major urinary protein 1, major urinary protein 10, major urinary protein 7, MUP 1, Mup-1, Mup1 major urinary protein 1, Mup10, Mup7, Mup-a, Up-1
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.