Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Netrin-G1a Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
| Antigen | Netrin-G1a |
|---|---|
| Clone | 229722 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
Netrin-G1a Monoclonal specifically detects Netrin-G1a in Mouse samples. It is validated for Western Blot.Specifications
| Netrin-G1a | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8R4G0 | |
| 22854 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Netrin-G1a His29-Gly513 Accession # Q8R4G0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Netrin-G1a in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant chicken (rch) Netrin-1, recombinant mouse Netrin-1, rchNetrin-2, or recombinant human Netrin-4 is observed. |
| 229722 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Lmnt1, NetrinG1a, NTG1, NTNG1 | |
| Ntng1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title