Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Nodal Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | Nodal |
---|---|
Clone | 209009 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Nodal Monoclonal specifically detects Nodal in Mouse samples. It is validated for Western Blot.Specifications
Nodal | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P43021 | |
4838 | |
E. coli-derived recombinant mouse Nodal His245-Leu354 Accession # P43021 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Nodal in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 25 - 50% cross-reactivity with recombinant Drosophila (rd) Dpp and no cross-reactivity with rmArtemin, rmCripto, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhLefty-A, rmLefty-1, rrMIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5. |
209009 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP-16, MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog | |
NODAL | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title