Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NPRA/NPR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3974SP
This item is not returnable.
View return policy
Description
NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Mouse samples. It is validated for Immunohistochemistry.Specifications
| NPRA/NPR1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P18293 | |
| NPR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 (predicted) Accession # P18293 | |
| 25 μg | |
| Primary | |
| Detects mouse NPRA/NPR1 in direct ELISAs. | |
| Mouse | |
| Purified |
| Immunohistochemistry | |
| 488620 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4881 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction