Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NPRA/NPR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | NPRA/NPR1 |
---|---|
Clone | 488620 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Mouse samples. It is validated for Immunohistochemistry.Specifications
NPRA/NPR1 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
P18293 | |
4881 | |
Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 (predicted) Accession # P18293 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse NPRA/NPR1 in direct ELISAs. |
488620 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr | |
NPR1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title