Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Persephin Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Persephin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Persephin Polyclonal specifically detects Persephin in Mouse samples. It is validated for Western Blot.Specifications
Persephin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
persephin, PSP, PSPN | |
PSPN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O70300.1 | |
5623 | |
E. coli-derived recombinant mouse Persephin Ala61-Gly156 Accession # O70300.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Persephin in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Persephin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title