Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PRDC/GREM2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2069SP
This item is not returnable.
View return policy
Description
PRDC/GREM2 Monoclonal specifically detects PRDC/GREM2 in Mouse samples. It is validated for Western Blot.Specifications
| PRDC/GREM2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O88273 | |
| GREM2 | |
| E. coli-derived recombinant mouse PRDC/GREM2 Arg22-Gln168 Accession # O88273 | |
| 25 μg | |
| Primary | |
| Detects mouse PRDC/GREM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) DAN, rmCerberus, rmGremlin, or rmCoco is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 377407 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CKTSF1B2, CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3, DAND3Prdc, GREM2, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64388 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction