Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PRDC/GREM2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | PRDC/GREM2 |
|---|---|
| Clone | 377407 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
PRDC/GREM2 Monoclonal specifically detects PRDC/GREM2 in Mouse samples. It is validated for Western Blot.Specifications
| PRDC/GREM2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| O88273 | |
| 64388 | |
| E. coli-derived recombinant mouse PRDC/GREM2 Arg22-Gln168 Accession # O88273 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse PRDC/GREM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) DAN, rmCerberus, rmGremlin, or rmCoco is observed. |
| 377407 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CKTSF1B2, CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3, DAND3Prdc, GREM2, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus | |
| GREM2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title