Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RANK/TNFRSF11A Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | RANK/TNFRSF11A |
---|---|
Clone | 100024 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Mouse samples. It is validated for Western Blot.Specifications
RANK/TNFRSF11A | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
TNFRSF11A | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
100024 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O35305 | |
8792 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse RANK/TNFRSF11A Leu29-Pro213 (Leu29Met) Accession # O35305 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse RANK/TNFRSF11A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) 4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmDR3, rmDR6, rmEDAR, rmFAS, rmGITR, recombinant human (rh) HVEM, rmLT beta R, rmNGF R, rmOPG, rmOX40, rhRELT, rmTROY, rmTNF RI, or rmTNF RII is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title