Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Serpin E1/PAI-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3828
This item is not returnable.
View return policy
Description
Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Mouse samples. It is validated for Western Blot.Specifications
Serpin E1/PAI-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
AAH54091 | |
SERPINE1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin E1/PAI-1 Leu24-Ser402 Accession # AAH54091 | |
100 μg | |
Primary | |
Detects mouse Serpin E1/PAI-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Serpin A1, A5, A6, B8, C1, D1, E2, F1, F2, I1, recombinant human Serpin A3, A4, B6, B9, G1, or I2 is observed. | |
Mouse | |
Purified |
Western Blot | |
367612 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5054 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction