Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Serpin E1/PAI-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Serpin E1/PAI-1 |
---|---|
Clone | 367612 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Serpin E1/PAI-1 Monoclonal specifically detects Serpin E1/PAI-1 in Mouse samples. It is validated for Western Blot.Specifications
Serpin E1/PAI-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
AAH54091 | |
5054 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin E1/PAI-1 Leu24-Ser402 Accession # AAH54091 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Serpin E1/PAI-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Serpin A1, A5, A6, B8, C1, D1, E2, F1, F2, I1, recombinant human Serpin A3, A4, B6, B9, G1, or I2 is observed. |
367612 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 | |
SERPINE1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title