Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Serpin I1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Serpin I1/Neuroserpin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Serpin I1/Neuroserpin Polyclonal specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for Western Blot.Specifications
Serpin I1/Neuroserpin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
5274 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin I1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Serpin I1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Serpin I1 is observed and less than 5% cross-reactivity with rhSerpin I2is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp781N13156, Neuroserpin, Peptidase inhibitor 12, PI12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1 | |
SERPINI1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title