Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Serum Albumin Antibody, Novus Biologicals™
SDP

Catalog No. NBP22651025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22651025 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22651025 Supplier Novus Biologicals Supplier No. NBP2265100.25MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mouse Serum Albumin Polyclonal specifically detects Mouse Serum Albumin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page.

Specifications

Antigen Mouse Serum Albumin
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:500 - 1:2500, SDS-Page
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias Alb1, Alb-1, albumin 1, BCL002
Gene Symbols Alb
Host Species Rabbit
Immunogen Anti-Mouse Serum Albumin Antibody was produced by repeated immunizations with mouse serum albumin.
Quantity 0.25 mg
Primary or Secondary Primary
Gene ID (Entrez) 11657
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.