Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Sonic Hedgehog/Shh C-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | Sonic Hedgehog/Shh |
---|---|
Clone | 55626 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot.Specifications
Sonic Hedgehog/Shh | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
SHH | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
55626 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q62226.1 | |
6469 | |
E. coli-derived recombinant mouse Sonic Hedgehog Cys199-Ser437 Accession # Q62226.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects recombinant mouse Shh C-terminal peptide in direct ELISAs and Western blots. Does not cross-react in direct ELISAs with rmDhh C- or N-terminal peptides, rmIhh C- or N-terminal peptides, rhShh N-terminal peptide, or rmShh N-terminal peptide. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title