Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Sulfamidase/SGSH Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2969
This item is not returnable.
View return policy
Description
Sulfamidase/SGSH Polyclonal specifically detects Sulfamidase/SGSH in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Sulfamidase/SGSH | |
Polyclonal | |
Unconjugated | |
NP_061292 | |
SGSH | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Sulfamidase/SGSH Arg23-Leu502 Accession # NP_061292 | |
100 μg | |
Primary | |
Detects mouse Sulfamidase/SGSH in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase | |
Goat | |
Affinity Purified | |
RUO | |
6448 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction